missing translation for 'onlineSavingsMsg'
Learn More

Methyl 11-Bromoundecanoate, TRC

Product Code. 30477911
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477911 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477911 Supplier Toronto Research Chemicals Supplier No. TRCM296585100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 11-Bromoundecanoate
CAS 6287-90-7
Molecular Formula C12H23O2Br
Quantity 100 mg
Synonym 11-bromo-Undecanoic acid methyl ester, 11-bromo-Hendecanoic acid methyl ester, 11-Bromoundecanoic acid methyl ester, NSC 12015
InChI Formula InChI=1S/C12H23BrO2/c1-15-12(14)10-8-6-4-2-3-5-7-9-11-13/h2-11H2,1H3
SMILES COC(=O)CCCCCCCCCCBr
IUPAC Name methyl 11-bromoundecanoate
Molecular Weight (g/mol) 279.21
Formula Weight 278.0881
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.