missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1H-indazole-6-carboxylate, 95%, Thermo Scientific™

Product Code. 15485717
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485717 1 g 1g
15495717 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15485717 Supplier Thermo Scientific Alfa Aesar Supplier No. H32933.03
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Methyl 1H-indazole-6-carboxylate is used in organic synthesis.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.  
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170487-40-8
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
MDL Number MFCD07371612
InChI Key TUSICEWIXLMXEY-UHFFFAOYSA-N
Synonym 1h-indazole-6-carboxylic acid methyl ester, methyl indazole-6-carboxylate, indazole-6-carboxylic acid methyl ester, 6-1h indazole carboxylic acid methyl ester, 1h-indazole-6-carboxylic acid, methyl ester, pubchem18064, methyl indazole-6carboxylate, ksc494o4p, methyl-1h-indazole-6-carboxylate, 2h-indazole-6-carboxylic acid methyl ester
PubChem CID 286535
IUPAC Name methyl 1H-indazole-6-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C=NN2

Specifications

Melting Point 142°C to 144°C
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 176.17
Percent Purity 95%
Chemical Name or Material Methyl 1H-indazole-6-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.