missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1H-pyrazole-3-carboxylate, 97%, Thermo Scientific Chemicals

Product Code. 15467077
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467077 1 g 1g
15477077 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467077 Supplier Thermo Scientific Alfa Aesar Supplier No. H34148.03
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
This item is not returnable. View return policy

Methyl 1H-pyrazole-3-carboxylate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 1H-pyrazole-3-carboxylate is used as a pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15366-34-4
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.115
MDL Number MFCD00649381
InChI Key ORUCTBNNYKZMSK-UHFFFAOYSA-N
Synonym methyl 1h-pyrazole-3-carboxylate, 1h-pyrazole-3-carboxylic acid methyl ester, methyl pyrazole-3-carboxylate, 2h-pyrazole-3-carboxylic acid methyl ester, 3-methoxycarbonylpyrazole, pyrazole-3-carboxylic acid methyl ester, methyl 2h-pyrazole-3-carboxylate, 1h-pyrazole-3-carboxylic acid, methyl ester, pubchem14332, 5-methoxycarbonylpyrazole
PubChem CID 565662
IUPAC Name methyl 1H-pyrazole-5-carboxylate
SMILES COC(=O)C1=CC=NN1

Specifications

Melting Point 141°C to 143°C
Quantity 1 g
Solubility Information Soluble in methanol.
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material Methyl 1H-pyrazole-3-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.