missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1H-pyrazole-3-carboxylate, Thermo Scientific™

Product Code. 10290189
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10290189 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10290189 Supplier Thermo Scientific Maybridge Supplier No. MAY00321DA
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 15366-34-4
CAS Min % 97.0
Packaging Amber Glass Bottle
Molecular Formula C5H6N2O2
Quantity 1 g
Synonym methyl 1h-pyrazole-3-carboxylate, 1h-pyrazole-3-carboxylic acid methyl ester, methyl pyrazole-3-carboxylate, 2h-pyrazole-3-carboxylic acid methyl ester, 3-methoxycarbonylpyrazole, pyrazole-3-carboxylic acid methyl ester, methyl 2h-pyrazole-3-carboxylate, 1h-pyrazole-3-carboxylic acid, methyl ester, pubchem14332, 5-methoxycarbonylpyrazole
InChI Key ORUCTBNNYKZMSK-UHFFFAOYSA-N
SMILES COC(=O)C1=CC=NN1
IUPAC Name methyl 1H-pyrazole-5-carboxylate
Molecular Weight (g/mol) 126.115
PubChem CID 565662
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material Methyl 1H-pyrazole-3-carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.