missing translation for 'onlineSavingsMsg'
Learn More

methyl 2-[1-methyl-3-(trifluoromethyl)-1h-pyrazol-5-yl]benzoate, 90%, Thermo Scientific™

Product Code. 10632315
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10632315 5 g Amber Glass Bottle 5g
10555754 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10632315 Supplier Thermo Scientific Maybridge Supplier No. CC62122DE
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 898289-62-8
Molecular Formula C13H11F3N2O2
Molecular Weight (g/mol) 284.238
MDL Number MFCD09879929
InChI Key VBXXEOGWKRJBBH-UHFFFAOYSA-N
Synonym methyl 2-1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl benzoate, methyl 2-2-methyl-5-trifluoromethyl pyrazol-3-yl benzoate, 2-2-methyl-5-trifluoromethyl-3-pyrazolyl benzoic acid methyl ester, 5-2-methoxycarbonyl phenyl-1-methyl-3-trifluoromethyl-1h-pyrazole, benzoic acid,2-1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl-, methyl ester
PubChem CID 24229663
IUPAC Name methyl 2-[2-methyl-5-(trifluoromethyl)pyrazol-3-yl]benzoate
SMILES CN1C(=CC(=N1)C(F)(F)F)C2=CC=CC=C2C(=O)OC

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 284.24
Percent Purity 90%
Chemical Name or Material Methyl 2-[1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.