missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate, 97%

Product Code. 15419487
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15419487 5 g 5g
15470086 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15419487 Supplier Thermo Scientific Alfa Aesar Supplier No. H52745.06
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 680-15-9 | C3H3F3O4S | 192.108 g/mol

Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate is a useful reagent for the trifluoromethylation of alkyl halides. It is also broadly applicable reagent for perfluoroalkylations for aryl iodides and bromides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate is a useful reagent for the trifluoromethylation of alkyl halides. It is also broadly applicable reagent for perfluoroalkylations for aryl iodides and bromides.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 680-15-9
Molecular Formula C3H3F3O4S
Molecular Weight (g/mol) 192.108
MDL Number MFCD00144316
InChI Key GQJCAQADCPTHKN-UHFFFAOYSA-N
Synonym methyl 2,2-difluoro-2-fluorosulfonyl acetate, methylfluorosulphonyldifluoroacetate, methyl difluoro fluorosulfonyl acetate, methyl fluorosulfonyldifluoroacetate, methyl fluorosulphonyl difluoro acetate, methyl fluorosulphonyldifluoroacetate, methyl 2-fluorosulfonyl difluoroacetate, fluorosulfonyldifluoroacetic acid methyl ester, 2,2-difluoro-2-fluorosulfonylacetic acid methyl ester, difluoro fluorosulfonyl acetic acid methyl ester
PubChem CID 547667
IUPAC Name methyl 2,2-difluoro-2-fluorosulfonylacetate
SMILES COC(=O)C(F)(F)S(=O)(=O)F

Specifications

Density 1.509
Boiling Point 117°C to 118°C
Flash Point 47°C (117°F)
Linear Formula FSO2CF2CO2CH3
Refractive Index 1.351
Quantity 5 g
UN Number UN2920
Beilstein 1812896
Formula Weight 192.11
Percent Purity 97%
Chemical Name or Material Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.