missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,3,4,6-Tetra-O-acetyl-Alpha-D-glucopyranoside, TRC

Product Code. 30478235
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478235 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478235 Supplier Toronto Research Chemicals Supplier No. TRCM326670500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 2,3,4,6-Tetra-O-acetyl-a-D-glucopyranoside
CAS 604-70-6
Molecular Formula C15 H22 O10
Quantity 500 mg
InChI Formula InChI=1S/C15H22O10/c1-7(16)21-6-11-12(22-8(2)17)13(23-9(3)18)14(24-10(4)19)15(20-5)25-11/h11-15H,6H2,1-5H3/t11-,12-,13+,14-,15+/m1/s1
SMILES CO[C@H]1O[C@H](COC(=O)C)[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]1OC(=O)C
IUPAC Name [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-methoxyoxan-2-yl]methyl acetate
Molecular Weight (g/mol) 362.33
Formula Weight 362.1213
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.