missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,5-dimethyl-1H-pyrrole-3-carboxylate, 95%, Thermo Scientific™

Product Code. p-2705154
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
This item is not returnable. View return policy

Product Code. 10570743

Brand: Thermo Scientific Maybridge SP00004DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 69687-80-5
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.181
MDL Number MFCD00203859
InChI Key OQWZEJIISPYZPW-UHFFFAOYSA-N
Synonym methyl 2,5-dimethylpyrrole-3-carboxylate, 1h-pyrrole-3-carboxylic acid, 2,5-dimethyl-, methyl ester, 2,5-dimethyl-1h-pyrrole-3-carboxylic acid methyl ester, maybridge1_008495, acmc-1b2uc, 2,5-dimethyl,4-carbomethoxy pyrrole, 2,5-dimethylpyrrole-3-carboxylic acid methyl ester, 1h-pyrrole-3-carboxylicacid,2,5-dimethyl-,methyl ester, 1h-pyrrole-3-carboxylicacid, 2,5-dimethyl-, methyl ester
PubChem CID 592729
IUPAC Name methyl 2,5-dimethyl-1H-pyrrole-3-carboxylate
SMILES CC1=CC(=C(N1)C)C(=O)OC

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 153.18
Percent Purity 95%
Chemical Name or Material Methyl 2, 5-dimethyl-1H-pyrrole-3-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.