missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloride, 97%, Thermo Scientific™

Product Code. 10360323
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10360323

Brand: Thermo Scientific Maybridge RH01792DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 5619-09-0
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C12H15ClN2O2
MDL Number MFCD00066133
Quantity 1 g
Synonym dl-tryptophan methyl ester hydrochloride, methyl 2-amino-3-1h-indol-3-yl propanoate hydrochloride, h-dl-trp-ome.hcl, dl-tryptophan methyl ester hcl, h-d-trp-ome hcl, methyl 2-amino-3-indol-3-ylpropanoate, chloride, h-dl-trp-ome hcl, h-dl-trp-ome-hcl, pubchem10907, acmc-209oxn
InChI Key XNFNGGQRDXFYMM-UHFFFAOYNA-N
SMILES Cl.COC(=O)C(N)CC1=CNC2=CC=CC=C12
IUPAC Name methyl 2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride
Molecular Weight (g/mol) 254.71
PubChem CID 2827264
Formula Weight 254.72
Percent Purity 97%
Chemical Name or Material Methyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.