missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate, 98%

Product Code. 15428195
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15428195 5 g 5g
15438195 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428195 Supplier Thermo Scientific Alfa Aesar Supplier No. H32755.06
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 108354-78-5 | C10H13NO2S | 211.279 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108354-78-5
Molecular Formula C10H13NO2S
Molecular Weight (g/mol) 211.279
MDL Number MFCD00130099
InChI Key DKYYKIHEIOOWRB-UHFFFAOYSA-N
Synonym methyl 2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylate, 2-amino-4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid methyl ester, 2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylic acid methyl ester, 2-amino-4,5,6,7-tetrahydro-benzo b thiophene-3, cbmicro_029799, methyl 2-azanyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate, methyl-2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylate, 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylic acid methyl ester, methyl 2-amino-4, 5, 6, 7-tetrahydro-1-benzothiophene-3-carboxylate, methyl 2-amino-4,5,6,7-tetrahydro-1-benzo b thiophene-3-carboxylate
PubChem CID 2756552
IUPAC Name methyl 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
SMILES COC(=O)C1=C(SC2=C1CCCC2)N

Specifications

Melting Point 126°C to 129°C
Quantity 5 g
Formula Weight 211.28
Percent Purity 98%
Chemical Name or Material Methyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.