missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate, 96%, Thermo Scientific™

Product Code. 15458685
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15458685 1 g 1g
15468685 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15458685

Brand: Thermo Scientific Alfa Aesar H33887.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 184174-81-0
Molecular Formula C11H15NO2S
Molecular Weight (g/mol) 225.306
MDL Number MFCD01114967
InChI Key BDPLKEMLVDUYRR-UHFFFAOYSA-N
Synonym methyl 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta b thiophene-3-carboxylate, methyl 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta-b thiophene-3-carboxylate, methyl 2-amino-4h,5h,6h,7h,8h-cyclohepta b thiophene-3-carboxylate, 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta b thiophene-3-carboxylic acid methyl ester, 5,6,7,8-tetrahydro-2-amino-4h-cyclohepta b thiophene-3-carboxylic acid methyl ester, methyl 2-amino-4,5,6,7,8-pentahydrocyclohepta 1,2-b thiophene-3-carboxylate, 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta b-thiophene-3-carboxylic acid methyl ester
PubChem CID 2756554
IUPAC Name methyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate
SMILES COC(=O)C1=C(SC2=C1CCCCC2)N

Specifications

Melting Point 95°C to 96°C
Quantity 1 g
Formula Weight 225.31
Percent Purity 96%
Chemical Name or Material Methyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.