missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Amino-5-bromobenzoate, TRC

Product Code. 30477985
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477985 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477985 Supplier Toronto Research Chemicals Supplier No. TRCM2857155G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 2-Amino-5-bromobenzoate
CAS 52727-57-8
Molecular Formula C8H8BrNO2
Quantity 5 g
Synonym 2-Amino-5-bromobenzoic Acid Methyl Ester, 5-Bromo-2-aminobenzoic Acid Methyl Ester, 5-Bromoanthranilic Acid Methyl Ester, Methyl 2-Amino-5-bromobenzoate, Methyl 5-Bromo-2-aminobenzoate, Methyl 5-Bromoanthranilate
InChI Formula InChI=1S/C8H8BrNO2/c1-12-8(11)6-4-5(9)2-3-7(6)10/h2-4H,10H2,1H3
SMILES COC(=O)c1cc(Br)ccc1N
IUPAC Name methyl 2-amino-5-bromobenzoate
Molecular Weight (g/mol) 230.06
Formula Weight 228.9738
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.