missing translation for 'onlineSavingsMsg'
Learn More

Methyl-2-chloropyrimidine-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10542585
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10542585 1 g Amber Glass Bottle 1g
10061094 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10542585 Supplier Thermo Scientific Maybridge Supplier No. MO07903DA
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 287714-35-6
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
MDL Number MFCD08690330
InChI Key VJOKXLBQCKCWLV-UHFFFAOYSA-N
Synonym 2-chloropyrimidine-5-carboxylic acid methyl ester, 5-pyrimidinecarboxylic acid, 2-chloro-, methyl ester, 2-chloro-5-pyrimidinecarboxylic acid methyl ester, pubchem9777, acmc-1cq94, 2-chloro-5-carbomethoxypyrimidine, methyl 2-chloro-5-pyrimidinecarboxylate, methyl-2-chloropyrimidine-5-carboxylate, methyl 2-chloranylpyrimidine-5-carboxylate, 2-chloro-5-methoxycarbonyl pyrimidine
PubChem CID 12627752
IUPAC Name methyl 2-chloropyrimidine-5-carboxylate
SMILES COC(=O)C1=CN=C(N=C1)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 172.57
Percent Purity 97%
Chemical Name or Material Methyl 2-chloropyrimidine-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.