missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(chlorosulfonyl)benzoate, 90%, Tech., Thermo Scientific™

Product Code. 10032830 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10032830 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10032830 Supplier Thermo Scientific Acros Supplier No. 224160250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26638-43-7
Molecular Formula C8H7ClO4S
Molecular Weight (g/mol) 234.65
MDL Number MFCD00009797
InChI Key HUNUAFNLLYVTQD-UHFFFAOYSA-N
Synonym methyl 2-chlorosulfonyl benzoate, benzoic acid, 2-chlorosulfonyl-, methyl ester, unii-c6hzm8x6pk, c6hzm8x6pk, methyl 2-chlorosulphonyl benzoate, pubchem14808, acmc-1cirj, methyl 2-chiorosulfonylbenzoate, methyl 2 chlorosulfonyl benzoate, methyl2-chlorosulfonyl benzoate
PubChem CID 117830
IUPAC Name methyl 2-(chlorosulfonyl)benzoate
SMILES COC(=O)C1=CC=CC=C1S(Cl)(=O)=O

Specifications

Melting Point 58°C to 64°C
Color Beige to White or Pink
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula ClSO2C6H4COOCH3
Quantity 25 g
Beilstein 11, 373
Formula Weight 234.66
Percent Purity ≥88% (ex Cl)
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Methyl 2-(chlorosulfonyl)benzoate, tech., 90%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 2-(chlorosulfonyl)benzoate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.