missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(chlorosulfonyl)benzoate, 94%

Product Code. 11400817
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11400817 5 g 5g
11410817 25 g 25g
11420817 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400817 Supplier Thermo Scientific Alfa Aesar Supplier No. B23211.06
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 26638-43-7 | C8H7ClO4S | 234.65 g/mol

Methyl 2-(chlorosulfonyl)benzoate can be used as intermediates in pharmaceuticals, saccharin, dyes, & pigments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-(chlorosulfonyl)benzoate can be used as intermediates in pharmaceuticals, saccharin, dyes, & pigments.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26638-43-7
Molecular Formula C8H7ClO4S
Molecular Weight (g/mol) 234.65
MDL Number MFCD00009797
InChI Key HUNUAFNLLYVTQD-UHFFFAOYSA-N
PubChem CID 117830
SMILES COC(=O)C1=CC=CC=C1S(Cl)(=O)=O

Specifications

Melting Point 59°C to 64°C
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Synonym methyl 2-chlorosulfonyl benzoate, benzoic acid, 2-chlorosulfonyl-, methyl ester, unii-c6hzm8x6pk, c6hzm8x6pk, methyl 2-chlorosulphonyl benzoate, pubchem14808, acmc-1cirj, methyl 2-chiorosulfonylbenzoate, methyl 2 chlorosulfonyl benzoate, methyl2-chlorosulfonyl benzoate
Solubility Information Reacts with water.
IUPAC Name methyl 2-chlorosulfonylbenzoate
Formula Weight 234.66
Percent Purity 94%
Chemical Name or Material Methyl 2-(chlorosulfonyl)benzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.