missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Cyclohexyl-2-hydroxyphenylacetate, TRC

Product Code. 30478184
Change view
Click to view available options
Quantity:
500 mg
10 g
Unit Size:
10g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478184 10 g 10g
30476346 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30478184 Supplier Toronto Research Chemicals Supplier No. TRCM29464510G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10399-13-0
Molecular Formula C15 H20 O3
Molecular Weight (g/mol) 248.32
Synonym Methyl (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetate, Phenylcyclohexylglycolic Acid Methyl Ester
IUPAC Name methyl 2-cyclohexyl-2-hydroxy-2-phenylacetate
SMILES COC(=O)C(O)(C1CCCCC1)c2ccccc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C15H20O3/c1-18-14(16)15(17,12-8-4-2-5-9-12)13-10-6-3-7-11-13/h2,4-5,8-9,13,17H,3,6-7,10-11H2,1H3
Formula Weight 248.1412
Chemical Name or Material Phenylcyclohexylglycolic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.