missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-cyclopentanonecarboxylate, 96%

Product Code. 10777772 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10777772 25 g Glass bottle 25g
10347321 5 g Glass bottle 5g
10249593 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10777772 Supplier Thermo Scientific Acros Supplier No. 342090250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10472-24-9 | C7H10O3 | 142.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10472-24-9
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.15
MDL Number MFCD00001411
InChI Key PZBBESSUKAHBHD-UHFFFAOYSA-N
Synonym methyl 2-oxocyclopentanecarboxylate, methyl 2-cyclopentanonecarboxylate, 2-carbmethoxycyclopentanone, methyl cyclopentanone-2-carboxylate, cyclopentanecarboxylic acid, 2-oxo-, methyl ester, 2-methoxy carbonyl cyclopentanone, 2-methoxycarbonylcyclopentanone, 2-oxocyclopentanecarboxylic acid methyl ester, cyclopentanone-2-carboxylic acid methyl ester, mehyl-2-oxocyclopentane
PubChem CID 66328
IUPAC Name methyl 2-oxocyclopentane-1-carboxylate
SMILES COC(=O)C1CCCC1=O

Specifications

Color Colorless to Yellow
Density 1.0540g/mL
Boiling Point 102°C to 104°C (11.0 mmHg)
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4550 to 1.457
Quantity 25 g
Beilstein 10, 597
Specific Gravity 1.054
Solubility Information Solubility in water: >100g/L. Other solubilities: miscible in ethanol
Formula Weight 142.15
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Methyl 2-cyclopentanonecarboxylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.