missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-fluoroacrylate, 95%, stab. with 1% BHT

Product Code. 15487868
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15487868 5 g 5g
15497868 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487868 Supplier Thermo Scientific Alfa Aesar Supplier No. H61343.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2343-89-7 | C4H5FO2 | 104.08 g/mol

Methyl 2-fluoroacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-fluoroacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2343-89-7
Molecular Formula C4H5FO2
Molecular Weight (g/mol) 104.08
MDL Number MFCD04039286
InChI Key ZTZJVAOTIOAZGZ-UHFFFAOYSA-N
Synonym methyl 2-fluoroacrylate, methyl-2-fluoroacrylate, 2-fluoroacrylic acid methyl ester, methyl fluoroacrylate, 2-propenoic acid, 2-fluoro-, methyl ester, methyl a-fluoroacrylate, pubchem12658, methyl alpha-fluoroacrylate, acmc-209g3b, ksc493a9d
PubChem CID 2782524
IUPAC Name methyl 2-fluoroprop-2-enoate
SMILES COC(=O)C(=C)F

Specifications

Linear Formula CH2=C(F)CO2CH3
Quantity 5 g
UN Number UN1993
Solubility Information Slightly soluble in water.
Formula Weight 104.08
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl 2-fluoroacrylate
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methyl 2-fluoroacrylate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H360-May damage fertility or the unborn child.
  • H372-Causes damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Methyl 2-fluoroacrylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.