Learn More
Methyl 2-fluoroacrylate, 95%, stab. with 1% BHT
Description
Methyl 2-fluoroacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl 2-fluoroacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Slightly soluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed.
Specifications
Specifications
| Linear Formula | CH2=C(F)CO2CH3 |
| Quantity | 25 g |
| UN Number | UN1993 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 104.08 |
| Percent Purity | 95% |
| Physical Form | Liquid |
| Chemical Name or Material | Methyl 2-fluoroacrylate |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.