missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-hexynoate, 98+%, Thermo Scientific™

Product Code. 10688234 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
Product Code. Quantity Packaging unitSize
10688234 25 g Glass bottle 25g
1 options
This item is not returnable. View return policy

Product Code. 10688234

Brand: Thermo Scientific Acros 382090250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 18937-79-6
MDL Number MFCD00041642
InChI Key DHZPZBFOJDXEHS-UHFFFAOYSA-N
Synonym methyl 2-hexynoate, 2-hexynoic acid methyl ester, 2-hexynoic acid, methyl ester, hex-2-ynoic acid methyl ester, methyl hexynoate, acmc-20akik, 2-hexynoicacid,methyl ester, 2-hexynoicacid, methyl ester
PubChem CID 87860
IUPAC Name methyl hex-2-ynoate
SMILES CCCC#CC(=O)OC

Specifications

Color Colorless to Yellow
Density 0.9430g/mL
Boiling Point 64.0°C to 66.0°C (10.0 mmHg)
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Molecular Formula C7H10O2
Linear Formula CH3(CH2)2C≡CCOOCH3
Refractive Index 1.4400 to 1.4420 (20°C, 589nm)
Quantity 25 g
Specific Gravity 0.943
Molecular Weight (g/mol) 126.15
Formula Weight 126.15
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Methyl 2-hexynoate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 2-hexynoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.