missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Hydroxy-3-methylbenzoate, TRC

Product Code. 30348662
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30348662 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348662 Supplier Toronto Research Chemicals Supplier No. TRCB5853351G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 2-Hydroxy-3-methylbenzoate
CAS 23287-26-5
Molecular Formula C9H10O3
Quantity 1 g
Synonym Benzoic acid, 2-hydroxy-3-methyl-, methyl ester, 2,3-Cresotic acid, methyl ester (6CI,7CI,8CI), 2-Hydroxy-3-methylbenzoic acid methyl ester, Levegal PT, Methyl 2-hydroxy-3-methylbenzoate, Methyl 3-methylsalicylate, Methyl o-cresotinate, NSC 165638, Methyl 2-Hydroxy-3-methylbenzoate
InChI Formula InChI=1S/C9H10O3/c1-6-4-3-5-7(8(6)10)9(11)12-2/h3-5,10H,1-2H3
SMILES COC(=O)c1cccc(C)c1O
IUPAC Name methyl 2-hydroxy-3-methylbenzoate
Molecular Weight (g/mol) 166.17
Formula Weight 166.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.