missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(hydroxymethyl)acrylate, 85%, stabilized with 0.1 wt.% MEHQ

Product Code. 11995081 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11995081 1 g 1g
11985081 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11995081 Supplier Thermo Scientific Acros Supplier No. 435900010
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 15484-46-5 | C5H8O3 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15484-46-5 , 150-19-6
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD12545894
InChI Key RFUCOAQWQVDBEU-UHFFFAOYSA-N
Synonym methyl 2-hydroxymethyl acrylate, methyl 2-hydroxymethyl prop-2-enoate, methyl2-hydroxymethyl acrylate, methyl-2-hydroxymethyl-acrylate, pubchem17373, methyl 2-hydroxymethylacrylate, methyl alpha-hydroxymethylacrylate, 2-hydroxymethylacrylate methyl ester, 2-methylolacrylic acid methyl ester, methyl alpha-hydroxymethyl acrylate
PubChem CID 11815329
IUPAC Name methyl 2-(hydroxymethyl)prop-2-enoate
SMILES COC(=O)C(=C)CO

Specifications

Color White
Density 1.1290g/mL
Boiling Point 71°C to 72°C (2.0 Torr)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 84% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.129
Stabilizer 0.1 wt% Monomethylether hydroquinone
Formula Weight 116.12
Percent Purity 85%
Physical Form Powder
Chemical Name or Material Methyl 2-(hydroxymethyl)acrylate, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 2-(hydroxymethyl)acrylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.