missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-methoxybenzoate, 98+%

Product Code. 11493278
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493278 100 g 100g
11483278 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493278 Supplier Thermo Scientific Alfa Aesar Supplier No. A17703.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 606-45-1 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 606-45-1
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00008423
InChI Key PFYHAAAQPNMZHO-UHFFFAOYSA-N
Synonym methyl o-anisate, dimethyl salicylate, o-anisic acid, methyl ester, methyl o-methoxybenzoate, o-methoxy methyl benzoate, o-methoxybenzoic acid methyl ester, methylsalicylate methyl ether, 2-methoxybenzoic acid methyl ester, benzoic acid, 2-methoxy-, methyl ester, 2-methoxybenzoic acid, methyl ester
PubChem CID 61151
IUPAC Name methyl 2-methoxybenzoate
SMILES COC1=CC=CC=C1C(=O)OC

Specifications

Density 1.16
Boiling Point 246°C to 248°C
Flash Point >110°C (230°F)
Odor Characteristic
Refractive Index 1.534
Quantity 100 g
Beilstein 2207356
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material Methyl 2-methoxybenzoate
compliance-icons
Product Identifier
  • Methyl 2-methoxybenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Methyl 2-methoxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.