missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Methyl-4-oxobutanoate (Technical Grade), TRC

Product Code. 30471231
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471231 2.5 mg 2.5mg
30467947 25 mg 25mg
30464679 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30471231 Supplier Toronto Research Chemicals Supplier No. TRCM3547382.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13865-21-9
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
IUPAC Name methyl 2-methyl-4-oxobutanoate
SMILES COC(=O)C(C)CC=O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C6H10O3/c1-5(3-4-7)6(8)9-2/h4-5H,3H2,1-2H3
Formula Weight 130.063
Chemical Name or Material Methyl 2-Methyl-4-oxobutanoate (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.