missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-methylbutyrate, 98%

Product Code. 11357985
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357985 100 g 100g
11347985 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357985 Supplier Thermo Scientific Alfa Aesar Supplier No. L08103.22
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 868-57-5 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868-57-5
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009335
InChI Key OCWLYWIFNDCWRZ-UHFFFAOYNA-N
Synonym methyl 2-methylbutyrate, butanoic acid, 2-methyl-, methyl ester, methyl 2-methyl butyrate, methyl s-2-methylbutanoate, 2-methylbutyric acid methyl ester, butyric acid, 2-methyl-, methyl ester, dl-2-methylbutyric acid methyl ester, methyl dl-2-methylbutyrate, methyl alpha-methylbutyrate, methyl alpha-methylbutanoate
PubChem CID 13357
IUPAC Name methyl 2-methylbutanoate
SMILES CCC(C)C(=O)OC

Specifications

Density 0.885
Boiling Point 114°C to 116°C
Flash Point 16°C (61°F)
Odor Fruit-like
Refractive Index 1.394
Quantity 100 g
UN Number UN3272
Beilstein 1720409
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material Methyl 2-methylbutyrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.