missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(methylsulfonyl)acetate, 97%, BLD Pharm™

Product Code. 31104370
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31104370 100 g
31095809 1 g
31104371 500 g
31093499 10 g
31103987 5 g
31106502 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31104370 Supplier BLD Pharm Supplier No. BD139508100g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 62020-09-1
Molecular Formula C4H8O4S
MDL Number MFCD00051842
Quantity 100 g
SMILES O=C(OC)CS(=O)(C)=O
Molecular Weight (g/mol) 152.17
Percent Purity 97%
Chemical Name or Material Methyl 2-(methylsulfonyl)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.