missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(methylsulfonylmethyl)acrylate, TRC

Product Code. 30482323
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482323 1 g 1g
30465439 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30482323 Supplier Toronto Research Chemicals Supplier No. TRCM3383781G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143224-94-6
Molecular Formula C6 H10 O4 S
Molecular Weight (g/mol) 178.206
IUPAC Name methyl 2-(methylsulfonylmethyl)prop-2-enoate
SMILES COC(=O)C(=C)CS(=O)(=O)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H10O4S/c1-5(6(7)10-2)4-11(3,8)9/h1,4H2,2-3H3
Formula Weight 178.03
Chemical Name or Material Methyl 2-(methylsulfonylmethyl)acrylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.