missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(methylthio)pyrimidine-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10132844
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10132844 250 mg Amber Glass Bottle 250mg
10470744 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10132844 Supplier Thermo Scientific Maybridge Supplier No. MO07902CB
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 38275-41-1
Molecular Formula C7H8N2O2S
Molecular Weight (g/mol) 184.21
MDL Number MFCD00194932
InChI Key PSYRMEZGAWNWHV-UHFFFAOYSA-N
PubChem CID 11194721
SMILES COC(=O)C1=CN=C(SC)N=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym methyl 2-methylthio pyrimidine-5-carboxylate, methyl 2-methylsulfanyl pyrimidine-5-carboxylate, methyl 2-methylthiopyrimidine-5-carboxylate, methyl 2-methylthio-5-pyrimidinecarboxylate, 2-methylthio-pyrimidine-5-carboxylic acid methyl ester, 5-pyrimidinecarboxylic acid, 2-methylthio-, methyl ester, methyl2-methylthio pyrimidine-5-carboxylate, methyl 2-methylthio-5-pyrimidinearboxylate, methyl-2-methylthio pyrimidine-5-carboxylate, methyl 2-methylsulphanyl pyrimidine-5-carboxylate
IUPAC Name methyl 2-(methylsulfanyl)pyrimidine-5-carboxylate
Formula Weight 184.22
Percent Purity 97%
Chemical Name or Material Methyl 2-(methylthio)pyrimidine-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.