missing translation for 'onlineSavingsMsg'
Learn More

Methyl-2-naphthalenemethylamine Hydrochloride, TRC

Product Code. 30477928
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477928 10 g 10g
30474347 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30477928 Supplier Toronto Research Chemicals Supplier No. TRCM32314510G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 381236-32-4
Molecular Formula C12H14ClN
Molecular Weight (g/mol) 207.7
Synonym N-Methyl-2-naphthalenemethanamine Hydrochloride, Methyl(naphthalen-2-ylmethyl)amine Hydrochloride, N-Methyl-N-(2-naphthylmethyl)amine, N-Methyl-N-[(naphthalen-2-yl)methyl]amine Hydrochloride
SMILES CNCC1=CC=C2C(C=CC=C2)=C1.Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H13N.ClH/c1-9-11(8-13)7-6-10-4-2-3-5-12(9)10;/h2-7H,8,13H2,1H3;1H
Chemical Name or Material Methyl-2-naphthalenemethylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.