missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-nonynoate, 98%, Thermo Scientific Chemicals

Product Code. 11499367
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499367 25 g 25g
11409377 100 g 100g
11419377 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499367 Supplier Thermo Scientific Alfa Aesar Supplier No. B21409.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-80-8
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.236
MDL Number MFCD00009547
InChI Key NTLJTUMJJWVCTL-UHFFFAOYSA-N
Synonym methyl 2-nonynoate, 2-nonynoic acid methyl ester, methyl octine carbonate, methyl octyne carbonate, 2-nonynoic acid, methyl ester, methyloctyne carboxylate, methyloctyne carbonate, methyl octin carbonate, octynecarboxylic acid, methyl ester, methyl octynecarboxylate
PubChem CID 8137
ChEBI CHEBI:51749
IUPAC Name methyl non-2-ynoate
SMILES CCCCCCC#CC(=O)OC

Specifications

Density 0.915
Boiling Point 121°C (20 mmHg)
Flash Point 104°C (219°F)
Odor Floral
Linear Formula CH3(CH2)5C≡CCO2CH3
Refractive Index 1.4485
Quantity 25 g
Beilstein 1759870
Formula Weight 168.24
Percent Purity 98%
Chemical Name or Material Methyl 2-nonynoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.