missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-phenylpyrimidine-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10440754
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10440754 1 g Amber Glass Bottle 1g
10386274 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10440754 Supplier Thermo Scientific Maybridge Supplier No. CC66322DA
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 64074-29-9
Molecular Formula C12H10N2O2
Molecular Weight (g/mol) 214.224
MDL Number MFCD06797254
InChI Key SOMJCOFUAUMQPG-UHFFFAOYSA-N
Synonym 2-phenylpyrimidine-5-carboxylic acid methyl ester, methyl2-phenylpyrimidine-5-carboxylate, 2-phenyl-pyrimidine-5-carboxylic acid methyl ester, 5-pyrimidinecarboxylicacid,2-phenyl-,methyl ester, 5-pyrimidinecarboxylicacid, 2-phenyl-, methyl ester, 5-pyrimidinecarboxylic acid, 2-phenyl-, methyl ester
PubChem CID 11206661
IUPAC Name methyl 2-phenylpyrimidine-5-carboxylate
SMILES COC(=O)C1=CN=C(N=C1)C2=CC=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 214.22
Percent Purity 97%
Chemical Name or Material Methyl 2-phenylpyrimidine-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.