missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-trifluoromethylsulfonyloxy-1-naphthoate, 98%, Thermo Scientific™

Product Code. 11466536
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11466536

Brand: Thermo Scientific Alfa Aesar B24793.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 200573-11-1
Molecular Formula C13H9F3O5S
Molecular Weight (g/mol) 334.27
MDL Number MFCD04039773
InChI Key BDXDWDCWSCPFEW-UHFFFAOYSA-N
Synonym methyl 2-trifluoromethylsulfonyloxy-1-naphthoate, methyl 2-trifluoromethanesulfonyloxy-1-naphthoate, methyl 2-trifluoromethyl sulfonyl oxy-1-naphthoate, methyl 2-trifluoromethanesulfonyloxy naphthalene-1-carboxylate, acmc-20aki5, methyl 2-trifluoromethylsulfonyloxy naphthalene-1-carboxylate, 1-carbomethoxy-2-trifluoromethylsulfonyloxy naphthalene, 2-trifluoromethylsulfonyloxy-1-naphthoic acid methyl ester, 2-trifluoromethylsulfonyloxy naphthalene-1-carboxylic acid methyl ester
PubChem CID 11034997
IUPAC Name methyl 2-(trifluoromethylsulfonyloxy)naphthalene-1-carboxylate
SMILES COC(=O)C1=C2C=CC=CC2=CC=C1OS(=O)(=O)C(F)(F)F

Specifications

Melting Point 41°C to 43°C
Quantity 5 g
Formula Weight 334.27
Percent Purity 98%
Chemical Name or Material Methyl 2-trifluoromethylsulfonyloxy-1-naphthoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.