missing translation for 'onlineSavingsMsg'
Learn More

Methyl (2S)-2-amino-2-(4-hydroxyphenyl)acetate Hydrochloride, TRC

Product Code. 30477900
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477900 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477900 Supplier Toronto Research Chemicals Supplier No. TRCM3390631G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl (2S)-Amino(4-hydroxyphenyl)ethanoate Hydrochloride
CAS 127369-30-6
Molecular Formula C9 H11 N O3 . Cl H
Quantity 1 g
Synonym Benzeneacetic acid, α-amino-4-hydroxy-, methyl ester, hydrochloride (1:1), (αS)-, Benzeneacetic acid, α-amino-4-hydroxy-, methyl ester, hydrochloride, (S)-, Benzeneacetic acid, α-amino-4-hydroxy-, methyl ester, hydrochloride, (αS)- (9CI), (S)-Methyl 2-amino-2-(4-hydroxyphenyl)acetate hydrochloride, Methyl (2S)-amino(4-hydroxyphenyl)ethanoate hydrochloride
InChI Formula InChI=1S/C9H11NO3.ClH/c1-13-9(12)8(10)6-2-4-7(11)5-3-6;/h2-5,8,11H,10H2,1H3;1H/t8-;/m0./s1
SMILES Cl.COC(=O)[C@@H](N)c1ccc(O)cc1
IUPAC Name methyl (2S)-2-amino-2-(4-hydroxyphenyl)acetate;hydrochloride
Molecular Weight (g/mol) 217.6495
Formula Weight 217.0506
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.