missing translation for 'onlineSavingsMsg'
Learn More

Methyl (2Z)-2-(acetylamino)-3-[1,1′-biphenyl]-4-yl-2-propenoate, TRC

Product Code. 30478088
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478088 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478088 Supplier Toronto Research Chemicals Supplier No. TRCM20238025MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl (2Z)-2-(acetylamino)-3-[1,1'-biphenyl]-4-yl-2-propenoate
CAS 556828-95-6
Molecular Formula C18H17NO3
Quantity 25 mg
Synonym Methyl (2Z)-2-(acetylamino)-3-[1,1′-biphenyl]-4-yl-2-propenoate, Methyl (2Z)-2-(acetylamino)-3-(biphenyl-4-yl)-2-propenoate, methyl 3-([1,1'-biphenyl]-4-yl)-2-acetamidoacrylate
InChI Formula InChI=1S/C18H17NO3/c1-13(20)19-17(18(21)22-2)12-14-8-10-16(11-9-14)15-6-4-3-5-7-15/h3-12H,1-2H3,(H,19,20)
SMILES O=C(OC)C(=CC=1C=CC(=CC1)C=2C=CC=CC2)NC(=O)C
IUPAC Name methyl 3-([1,1'-biphenyl]-4-yl)-2-acetamidoacrylate
Molecular Weight (g/mol) 295.33
Formula Weight 295.12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.