missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(2-Ethyl-4-oxo-1-phenyl-1,3,8-triazaspiro[4.5]dec-2-en-8-yl)propanoate, TRC

Product Code. 30477812
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477812 10 mg 10mg
30471522 2.5 mg 2.5mg
30475884 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477812 Supplier Toronto Research Chemicals Supplier No. TRCM31675510MG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2518276-69-0
Molecular Formula C19H25N3O3
Molecular Weight (g/mol) 343.43
Synonym Remifentanil EP Impurity L
IUPAC Name methyl 3-(2-ethyl-4-oxo-1-phenyl-1,3,8-triazaspiro[4.5]dec-2-en-8-yl)propanoate
SMILES CCC1=NC(=O)C2(CCN(CCC(=O)OC)CC2)N1c3ccccc3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H25N3O3/c1-3-16-20-18(24)19(22(16)15-7-5-4-6-8-15)10-13-21(14-11-19)12-9-17(23)25-2/h4-8H,3,9-14H2,1-2H3
Formula Weight 343.1896
Chemical Name or Material Methyl 3-(2-Ethyl-4-oxo-1-phenyl-1,3,8-triazaspiro[4.5]dec-2-en-8-yl)propanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.