missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(2-morpholin-4-ylethoxy)benzoate, 97%, Thermo Scientific™

Product Code. 10403793
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10403793 1 g 1g
10497934 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10403793 Supplier Thermo Scientific Maybridge Supplier No. CC42022DA
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 249937-00-6
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD08060518
InChI Key VOYSHDFODOJUMP-UHFFFAOYSA-N
Synonym methyl 3-2-morpholin-4-ylethoxy benzoate, methyl 3-2-morpholin-4-yl ethoxy benzoate, methyl 3-2-morpholinoethoxy benzoate, methyl 3-2-morpholin4-ylethoxy benzoate, 3-2-morpholin-4-yl-ethoxy-benzoic acid methyl ester, benzoic acid,3-2-4-morpholinyl ethoxy-, methyl ester
PubChem CID 7164628
IUPAC Name methyl 3-(2-morpholin-4-ylethoxy)benzoate
SMILES COC(=O)C1=CC(OCCN2CCOCC2)=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 265.31
Percent Purity 97%
Chemical Name or Material Methyl 3-(2-morpholin-4-ylethoxy)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.