missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(2-thienyl)acrylate, 97%, Thermo Scientific™

Product Code. 10597284
Change view
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10597284 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10597284 Supplier Thermo Scientific Maybridge Supplier No. KM05235EB
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 20883-96-9
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H8O2S
MDL Number MFCD00173772
Quantity 25 g
Synonym methyl 3-2-thienyl acrylate, methyl 3-thien-2-yl acrylate, methyl 3-thien-2-yl-acrylate, methyl e-3-thiophen-2-ylprop-2-enoate, methyl 2e-3-thiophen-2-yl prop-2-enoate, methyl 3-thiophen-2-yl prop-2-enoate, thiophene-2-acrylic acid methyl ester, e-methyl 3-thiophen-2-yl acrylate, methyl 2e-3-2-thienyl-2-propenoate, methyl 2e-3-2-thienyl prop-2-enoate
InChI Key HKVOGMDMMCLQFJ-SNAWJCMRSA-N
SMILES COC(=O)C=CC1=CC=CS1
IUPAC Name methyl (E)-3-thiophen-2-ylprop-2-enoate
Molecular Weight (g/mol) 168.21
PubChem CID 5709013
Formula Weight 168.22
Percent Purity 97%
Chemical Name or Material Methyl 3-(2-thienyl)acrylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.