missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-[3-(dimethylamino)propoxy]benzoate, 97%, Thermo Scientific™

Product Code. 11396835
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11396835 5 g Amber Glass Bottle 5g
10422504 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11396835 Supplier Thermo Scientific Maybridge Supplier No. CC59422DE
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 921938-75-2
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.299
MDL Number MFCD09702393
InChI Key WGAZXKVAWOAUOI-UHFFFAOYSA-N
Synonym methyl 3-3-dimethylamino propoxy benzoate
PubChem CID 24229605
IUPAC Name methyl 3-[3-(dimethylamino)propoxy]benzoate
SMILES CN(C)CCCOC1=CC=CC(=C1)C(=O)OC

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 237.3
Percent Purity 97%
Chemical Name or Material Methyl 3-[3-(dimethylamino)propoxy]benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.