missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(3-pyridyl)propionate, 98%

Product Code. 11353606
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11353606 1 g 1g
11363606 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11353606

Brand: Thermo Scientific Alfa Aesar L14590.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 84199-98-4 | C9H11NO2 | 165.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84199-98-4
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD01075660
InChI Key FQRRQPZYIYAPAV-UHFFFAOYSA-N
Synonym methyl 3-3-pyridyl propionate, methyl 3-pyridin-3-yl propanoate, methyl 3-3-pyridyl propanoate, methyl 3-3-pyridinyl propanoate, 3-pyridinepropanoicacid, methyl ester, 3-3-pyridyl-propionic acid methyl ester
PubChem CID 10329646
IUPAC Name methyl 3-pyridin-3-ylpropanoate
SMILES COC(=O)CCC1=CN=CC=C1

Specifications

Boiling Point 134°C (12 mmHg)
Refractive Index 1.501
Quantity 1 g
Beilstein 132852
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material Methyl 3-(3-pyridyl)propionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.