missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4-diaminobenzoate, 98%

Product Code. 11497616
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11497616 25 g 25g
11487616 5 g 5g
11407626 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 11497616

Brand: Thermo Scientific Alfa Aesar A14656.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 36692-49-6 | C8H10N2O2 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36692-49-6
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00017098
InChI Key IOPLHGOSNCJOOO-UHFFFAOYSA-N
Synonym 3,4-diaminobenzoic acid methyl ester, methyl3,4-diaminobenzoate, methyl-3,4-diaminobenzoate, benzoic acid, 3,4-diamino-, methyl ester, 3,4-diamino-benzoic acid methyl ester, methyl 3,4-diaminobenzenecarboxylate, pubchem4610, acmc-1bn7s, methyl 3,4-diamino-benzoate, methyl-3,4-diamino-benzoate
PubChem CID 135524
IUPAC Name methyl 3,4-diaminobenzoate
SMILES COC(=O)C1=CC=C(N)C(N)=C1

Specifications

Melting Point 105°C to 108°C
Quantity 25 g
Beilstein 2091675
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material Methyl 3,4-diaminobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.