missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4-dichlorophenylacetate, 99%, Thermo Scientific™

Product Code. 10068811 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10068811 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10068811 Supplier Thermo Scientific Acros Supplier No. 351080250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6725-44-6
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H8Cl2O2
Refractive Index 1.5400 to 1.5420
MDL Number MFCD00800680
Quantity 25 g
Synonym methyl 2-3,4-dichlorophenyl acetate, methyl 3,4-dichlorophenylacetate, 3,4-dichlorophenylacetic acid methyl ester, 3,4-dichloro-phenyl-acetic acid methyl ester, benzeneacetic acid, 3,4-dichloro-, methyl ester, pubchem20251, acmc-1bhwn, methyl 3,4-dichloro-phenylacetate, methyl 2-3,4-dichlorophenyl ethanoate, 3,4 dichlorophenyl acetic acid methyl ester
InChI Key FWVOAIHAIWHHDM-UHFFFAOYSA-N
SMILES COC(=O)CC1=CC(=C(C=C1)Cl)Cl
IUPAC Name methyl 2-(3,4-dichlorophenyl)acetate
Molecular Weight (g/mol) 219.07
PubChem CID 1268257
Formula Weight 219.07
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material Methyl 3, 4-dichlorophenylacetate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 3,4-dichlorophenylacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.