missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(4-hydroxyphenyl)propionate, 98%

Product Code. 10475672 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10475672 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10475672 Supplier Thermo Scientific Acros Supplier No. 369430050
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 5597-50-2 | C10H12O3 | 180.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5597-50-2
Melting Point 37.0°C to 41.0°C
Color White
Boiling Point 108.0°C (11.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C10H12O3
Linear Formula HOC6H4CH2CH2CO2CH3
MDL Number MFCD00071577
Quantity 5 g
Synonym methyl 3-4-hydroxyphenyl propionate, methyl 3-4-hydroxyphenyl propanoate, benzenepropanoic acid, 4-hydroxy-, methyl ester, unii-wat13au7xb, methyl3-4-hydroxyphenyl propionate, wat13au7xb, hydrocinnamic acid, p-hydroxy-, methyl ester, 3-4-hydroxyphenyl propionic acid methyl ester, methyl 3-p-hydroxyphenyl propionate, 3-4'-hydroxyphenyl-propionic acid-ome
InChI Key XRAMJHXWXCMGJM-UHFFFAOYSA-N
SMILES COC(=O)CCC1=CC=C(C=C1)O
IUPAC Name methyl 3-(4-hydroxyphenyl)propanoate
Molecular Weight (g/mol) 180.2
PubChem CID 79706
Formula Weight 180.2
Percent Purity 98%
Physical Form Low Melting Mass
Chemical Name or Material Methyl 3-(4-hydroxyphenyl)propionate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 3-(4-hydroxyphenyl)propionate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.