missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-dimethylbenzoate, 98%

Product Code. 11455013
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455013 25 g 25g
11445013 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455013 Supplier Thermo Scientific Alfa Aesar Supplier No. A12287.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 25081-39-4 | C10H12O2 | 164.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25081-39-4
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00077971
InChI Key PEVXENGLERTHJE-UHFFFAOYSA-N
PubChem CID 32786
IUPAC Name methyl 3,5-dimethylbenzoate
SMILES CC1=CC(=CC(=C1)C(=O)OC)C

Specifications

Melting Point 31°C to 33°C
Density 1.027
Quantity 25 g
Synonym benzoic acid, 3,5-dimethyl-, methyl ester, 3,5-dimethylbenzoic acid methyl ester, acmc-1cmfo, methyl-3,5-dimethylbenzoate, methyl-3,5-dimethyl-benzoate, 3,5-dimethylbenzoic acid methyl, methyl 3,5-dimethylbenzoate, 3,5-ch3 2-c6h3-cooch3, benzoic acid,3,5-dimethyl-, methyl ester, 3,5-dimethyl-benzoic acid methyl ester
Boiling Point 239°C to 240°C
Formula Weight 164.21
Percent Purity 98%
Chemical Name or Material Methyl 3,5-dimethylbenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.