missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-4-methylthiophene-2-carboxylate, 98%, Thermo Scientific™

Product Code. 11449362
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449362 100 g 100g
11429362 5 g 5g
11439362 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449362 Supplier Thermo Scientific Alfa Aesar Supplier No. A10115.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85006-31-1
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.21
MDL Number MFCD00051822
InChI Key YICRPERKKBDRSP-UHFFFAOYSA-N
Synonym methyl3-amino-4-methylthiophene-2-carboxylate, 3-amino-4-methylthiophene-2-carboxylic acid methyl ester, unii-3la24473as, 3-amino-4-methyl-2-thiophenecarboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-amino-4-methyl-, methyl ester, methyl 3-amino-4-methyl-2-thenoate, methyl 3-amino-4-methyl-2-thiophenecarboxylate, methyl-3-amino-4-methylthiophene-2-carboxylate, methyl 3-amino-4-methyl-thiophene-2-carboxylate, 3-amino-4-methyl-thiophene-2-carboxylic acid methyl ester
PubChem CID 123584
IUPAC Name methyl 3-amino-4-methylthiophene-2-carboxylate
SMILES COC(=O)C1=C(N)C(C)=CS1

Specifications

Melting Point 84°C to 88°C
Flash Point >100°C (212°F)
Quantity 100 g
Beilstein 973196
Formula Weight 171.22
Percent Purity 98%
Chemical Name or Material Methyl 3-amino-4-methylthiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.