missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-5-methylthiophene-2-carboxylate, 97%

Product Code. 15458476
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458476 5 g 5g
15468476 250 mg 250mg
15448476 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15458476 Supplier Thermo Scientific Alfa Aesar Supplier No. H63719.06
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 76575-71-8 | C7H9NO2S | 171.21 g/mol

Methyl 3-amino-5-methylthiophene-2-carboxylate is employed as a intermediate for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-amino-5-methylthiophene-2-carboxylate is employed as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76575-71-8
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.21
MDL Number MFCD00130095
InChI Key FVKMOPIFLCMZMI-UHFFFAOYSA-N
Synonym 3-amino-5-methyl-thiophene-2-carboxylic acid methyl ester, methyl 3-amino-5-methyl thiophene-2-carboxylate, 3-amino-5-methylthiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-amino-5-methyl-, methyl ester, methyl3-amino-5-methylthiophene-2-carboxylate, 3-amino-5-methylthiophene-2-carboxylate, 3-amino-5-methyl-thiophene-2-carboxylic acid, methyl 3-amino-5-methylthiophen-2-carboxylate, methyl-3-amino-5-methylthiophene-2-carboxylate, 3-amino-5-methyl-2-thiophene-carboxylic acid, methyl ester
PubChem CID 818952
IUPAC Name methyl 3-amino-5-methylthiophene-2-carboxylate
SMILES COC(=O)C1=C(N)C=C(C)S1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 171.22
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 3-amino-5-methylthiophene-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.