missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-aminocrotonate, 97%

Product Code. 10091680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10091680 100 g Plastic bottle 100g
10317670 5 g Glass bottle 5g
10377710 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10091680 Supplier Thermo Scientific Acros Supplier No. 156791000
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14205-39-1 | C5H9NO2 | 115.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14205-39-1
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD00008072,MFCD00008072,MFCD00008072
InChI Key XKORCTIIRYKLLG-ONEGZZNKSA-N
Synonym methyl 3-aminocrotonate, methyl 3-aminobut-2-enoate, methyl 2z-3-aminobut-2-enoate, z-methyl 3-aminobut-2-enoate, 3-aminocrotonic acid methyl ester, methyl beta-aminocrotonate, methyl z-3-aminobut-2-enoate, methyl 2z-3-amino-2-butenoate, beta-aminocrotonic acid methyl ester, 3-amino-2-butenoic acid methyl ester
PubChem CID 643918
IUPAC Name methyl (Z)-3-aminobut-2-enoate
SMILES COC(=O)\C=C(/C)N

Specifications

Melting Point 81.0°C to 83.0°C
Color White to Yellow
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Linear Formula CH3C(NH2)=CHCO2CH3
Quantity 100 g
Beilstein 03, 632
Solubility Information Solubility in water: insoluble in water. Other solubilities: solubility in chloroform >500g/L (20°C),solubility in acetone >500g/L (20°C)
Formula Weight 115.13
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-aminocrotonate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 3-aminocrotonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.