missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-aminocrotonate, 97%

Product Code. 11491487
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491487 100 g 100g
11401497 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491487 Supplier Thermo Scientific Alfa Aesar Supplier No. B24953.22
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14205-39-1 | C5H9NO2 | 115.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14205-39-1
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD00008072,MFCD00008072,MFCD00008072
InChI Key XKORCTIIRYKLLG-ONEGZZNKSA-N
Synonym methyl 3-aminocrotonate, methyl 3-aminobut-2-enoate, methyl 2z-3-aminobut-2-enoate, z-methyl 3-aminobut-2-enoate, 3-aminocrotonic acid methyl ester, methyl beta-aminocrotonate, methyl z-3-aminobut-2-enoate, methyl 2z-3-amino-2-butenoate, beta-aminocrotonic acid methyl ester, 3-amino-2-butenoic acid methyl ester
PubChem CID 643918
IUPAC Name methyl (Z)-3-aminobut-2-enoate
SMILES COC(=O)\C=C(/C)N

Specifications

Melting Point 81°C to 85°C
Boiling Point 122°C (42 mmHg)
Flash Point 91°C (195°F)
Quantity 100 g
Beilstein 956592
Sensitivity Hygroscopic
Formula Weight 115.13
Percent Purity 97%
Chemical Name or Material Methyl 3-aminocrotonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.