missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-aminothiophene-4-carboxylate hydrochloride, 97+%

Product Code. 11315657
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11315657 5 g 5g
11410580 0.25 g 0.25g
11305657 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11315657 Supplier Thermo Scientific Alfa Aesar Supplier No. 043384.06
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 39978-14-8 | C6H7NO2S | 157.19 g/mol

Methyl 3-aminothiophene-4-carboxylate hydrochloride is used in the preparation of ethyl (4-methoxycarbonyl)thiophene-3-carbamate by reacting with carbonochloridic acid ethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39978-14-8
Molecular Formula C6H7NO2S
Molecular Weight (g/mol) 157.19
MDL Number MFCD00068149
InChI Key BUFZZXCVOFBHLS-UHFFFAOYSA-N
Synonym methyl 4-aminothiophene-3-carboxylate hydrochloride, methyl 3-aminothiophene-4-carboxylate hydrochloride, 3-carbomethoxy-4-aminothiophene hydrochloride, acmc-209zkz, methyl 4-aminothiophene-3-carboxylate, chloride, 3-amino-4-methoxycarbonylthiophene hydrochloride, 4-amino-3-methoxycarbonylthiophene hydrochloride, methyl3-aminothiophene-4-carboxylatehydrochloride, 4-amino-3-methoxycarbonyl thiophene hydrochloride, methyl 4-aminothiophen-3-carboxylate hydrochloride
PubChem CID 2777609
SMILES COC(=O)C1=CSC=C1N

Specifications

Melting Point 199°C to 200°C (decomposition)
Color Brown
Quantity 5 g
Sensitivity Hygroscopic
IUPAC Name methyl 4-aminothiophene-3-carboxylate;hydrochloride
Formula Weight 193.65
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material Methyl 3-aminothiophene-4-carboxylate hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.