missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate, 95%, Thermo Scientific Chemicals

Product Code. 15429065
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15429065 1 g 1g
15439065 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15429065 Supplier Thermo Scientific Alfa Aesar Supplier No. H35994.03
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 342613-63-2 | C9H7BrN2O2 | 255.071 g/mol

Methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 342613-63-2
Molecular Formula C9H7BrN2O2
Molecular Weight (g/mol) 255.071
MDL Number MFCD10566741
InChI Key VFFFBWRUZMAHFS-UHFFFAOYSA-N
Synonym methyl 3-bromoimidazo 1,2-a pyridine-7-carboxylate, 3-bromo-7-methoxycarbonyl imidazo 1,2-a pyridine, methyl 3-bromoh-imidazo 1,2-a pyridine-7-carboxylate, 3-bromoimidazo 1,2-a pyridine-7-carboxylic acid methyl ester, pubchem23890, methyl3-bromoimidazo 1,2-a pyridine-7-carboxylate, 3-bromo-imidazo 1,2-a pyridine-7-carboxylic acid methyl ester, methyl 3-bromo-5h-imidazo 2,1-b 1,3 oxazine-7-carboxylate, 3-bromo-imidazo 1,2-alpha pyridine-7-carboxylic acid methyl ester, 3-bromoimidazo 1,2-alpha pyridine-7-carboxylic acid methyl ester
PubChem CID 45790121
IUPAC Name methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate
SMILES COC(=O)C1=CC2=NC=C(N2C=C1)Br

Specifications

Melting Point 156°C to 157°C
Quantity 1 g
Formula Weight 255.07
Percent Purity 95%
Chemical Name or Material Methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.