missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(chlorosulfonyl)-4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate, 95%, Thermo Scientific™

Product Code. 10601313
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10601313 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10601313 Supplier Thermo Scientific Maybridge Supplier No. SEW03400DA
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 306935-98-8
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C13H8ClF3O4S2
MDL Number MFCD02090166
Quantity 1 g
Synonym methyl 3-chlorosulfonyl-4-phenyl-5-trifluoromethyl thiophene-2-carboxylate, 2-thiophenecarboxylicacid, 3-chlorosulfonyl-4-phenyl-5-trifluoromethyl-, methyl ester, 3-chlorosulfonyl-4-phenyl-5-trifluoromethyl-thiophene-2-carboxylic acid methyl ester, methyl 3-chlorosulphonyl-4-phenyl-5-trifluoromethyl thiophene-2-carboxylate, 2-methoxycarbonyl-4-phenyl-5-trifluoromethyl thiophene-3-sulphonyl chloride, 3-chlorosulfonyl-4-phenyl-5-trifluoromethyl-2-thiophenecarboxylic acid methyl ester, methyl 3-chloranylsulfonyl-4-phenyl-5-trifluoromethyl thiophene-2-carboxylate
InChI Key QTEYQFWTRAYGTC-UHFFFAOYSA-N
SMILES COC(=O)C1=C(C(=C(S1)C(F)(F)F)C1=CC=CC=C1)S(Cl)(=O)=O
IUPAC Name methyl 3-chlorosulfonyl-4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
Molecular Weight (g/mol) 384.77
PubChem CID 2778708
Formula Weight 384.78
Percent Purity 95%
Chemical Name or Material Methyl 3-(chlorosulfonyl)-4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.